accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16343-08-1

IUPAC Name: hexylboronic acid
Synonyms: hexylboronic acid
Molecular Weight (g/mol): 129.99
Molecular Formula: C6H15BO2
InChi Key: CXSYDLCMCLCOCA-UHFFFAOYSA-N
SMILES: CCCCCCB(O)O
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)