Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. H09135G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0913-5G 5 g
H09131G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H0913-5G Supplier TCI America Supplier No. H09135G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 16343-08-1
Molecular Formula C6H15BO2
MDL Number MFCD01074641
InChI Key CXSYDLCMCLCOCA-UHFFFAOYSA-N
PubChem CID 351064
IUPAC Name hexylboronic acid

Specifications

Color White-Yellow
Quantity 5 g
Synonym n-hexylboronic acid, 1-hexaneboronic acid, boronic acid, b-hexyl, 1-hexylboronic acid, n-hexaneboronic acid
SMILES CCCCCCB(O)O
Molecular Weight (g/mol) 129.99
Formula Weight 129.99
Physical Form Crystalline Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.