accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16347-60-7

IUPAC Name: 3-phenyl-3,4-dihydroquinazolin-4-one
Synonyms: 3-phenylquinazolin-4-one
Molecular Weight (g/mol): 222.25
Molecular Formula: C14H10N2O
InChi Key: WAIHFZPSLVDBRV-UHFFFAOYSA-N
SMILES: O=C1N(C=NC2=CC=CC=C12)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results