accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16355-92-3

IUPAC Name: 1,10-diiododecane
Synonyms: 1,10-diiododecane
Molecular Weight (g/mol): 394.08
Molecular Formula: C10H20I2
InChi Key: CKJCTZAIDVFHCX-UHFFFAOYSA-N
SMILES: ICCCCCCCCCCI
Molecular Weight (g/mol) 
  • (1)
  • (5)
Percent Purity 
  • (1)
  • (1)
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (4)
  • (1)
Physical Form 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (4)
  • (1)
  • (5)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)