Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,10-Diiododecane, 97%, Thermo Scientific™

Catalog No. AC249620250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC249620250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC249620250 Supplier Thermo Scientific Chemicals Supplier No. 249620250

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16355-92-3
Molecular Formula C10H20I2
Molecular Weight (g/mol) 394.079
InChI Key CKJCTZAIDVFHCX-UHFFFAOYSA-N
Synonym decamethylene diiodide, decane, 1,10-diiodo, 1,10-diiododecan, pubchem3844, acmc-1btxk, decane,1,10-diiodo, 1,10-diiododecane
PubChem CID 85378
IUPAC Name 1,10-diiododecane
SMILES C(CCCCCI)CCCCI

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 28°C to 33°C
Color Brown to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Linear Formula I(CH2)10I
Quantity 25 g
Beilstein 01, 168
Formula Weight 394.07
Percent Purity ≥96.0%
Physical Form Crystalline Solid
Chemical Name or Material 1, 10-Diiododecane, 97%
Show More Show Less
missing translation for 'healthHazard1' LIGHT SENSITIVE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.