accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16361-24-3

IUPAC Name: 5-chloro-1-benzothiophene-3-carboxylic acid
Synonyms: 5-chloro-1-benzothiophene-3-carboxylic acid
Molecular Weight (g/mol): 212.65
Molecular Formula: C9H5ClO2S
InChi Key: RTBIQTHYQWFIAO-UHFFFAOYSA-N
SMILES: OC(=O)C1=CSC2=CC=C(Cl)C=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results