Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Chloro-benzo[b]thiophene-3-carboxylic acid, 97%, Thermo Scientific™

Catalog No. CC00115DE
Change view
Click to view available options
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
CC00115DE 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC00115DE Supplier Thermo Scientific Chemicals Supplier No. CC00115DE

missing translation for 'validMainframeMsg'

Specifications

CAS 16361-24-3
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C9H5ClO2S
Quantity 5g
MDL Number MFCD00052301
Synonym 5-chloro-benzo b thiophene-3-carboxylic acid, 5-chlorobenzo b thiophene-3-carboxylic acid, 5-chloranyl-1-benzothiophene-3-carboxylic acid, benzo b thiophene-3-carboxylicacid, 5-chloro
InChI Key RTBIQTHYQWFIAO-UHFFFAOYSA-N
SMILES OC(=O)C1=CSC2=CC=C(Cl)C=C12
IUPAC Name 5-chloro-1-benzothiophene-3-carboxylic acid
Molecular Weight (g/mol) 212.65
PubChem CID 2794574
Formula Weight 212.66
Percent Purity 97%
Chemical Name or Material 5-Chloro-benzo[b]thiophene-3-carboxylic acid
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.