accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2FFOH

CAS RN 163733-99-1

IUPAC Name: 4-amino-2,3-difluorophenol
Synonyms: 4-amino-2,3-difluorophenol
Molecular Weight (g/mol): 145.11
Molecular Formula: C6H5F2NO
InChi Key: LYFMJLYBTMEYDV-UHFFFAOYSA-N
SMILES: NC1=C(F)C(F)=C(O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results