Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-2,3-difluorophenol, 98%, Thermo Scientific™

Catalog No. AAH31555MD
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH31555MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH31555MD Supplier Thermo Scientific Chemicals Supplier No. H31555MD

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 163733-99-1
Molecular Formula C6H5F2NO
Molecular Weight (g/mol) 145.11
MDL Number MFCD03094494
InChI Key LYFMJLYBTMEYDV-UHFFFAOYSA-N
Synonym 4-amino-2,3-difluoro-phenol, 2,3-difluoro-4-hydroxyaniline, phenol, 4-amino-2,3-difluoro, phenol, 4-amino-2,3-difluoro-9ci, pubchem3541, 4-amino-2,3difluoro-phenol, ksc495e3n, 4-hydroxy-2,3-difluoroaniline
PubChem CID 2778766
IUPAC Name 4-amino-2,3-difluorophenol
SMILES NC1=C(F)C(F)=C(O)C=C1

Specifications

Melting Point ∽153°C (decomposition)
Quantity 250 mg
Formula Weight 145.11
Percent Purity 98%
Chemical Name or Material 4-Amino-2,3-difluorophenol
Hazard Category H302+H312-H315-H319-H335
Hazard Statement GHS H Statement
H301-H311-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.