accessibility menu, dialog, popup

UserName

Viewing profile as user:
N(E)

CAS RN 16375-56-7

IUPAC Name: 4-[(1E)-2-(naphthalen-1-yl)ethenyl]pyridine
Synonyms: naphthylvinylpyridine
Molecular Weight (g/mol): 231.3
Molecular Formula: C17H13N
InChi Key: FLKDRTOVVLNOLV-CMDGGOBGSA-N
SMILES: C(=C/C1=CC=CC2=CC=CC=C12)\C1=CC=NC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results