accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 163765-44-4

IUPAC Name: tert-butyl (3R)-3-methylpiperazine-1-carboxylate
Synonyms: tert-butyl (3R)-3-methylpiperazine-1-carboxylate
Molecular Weight (g/mol): 200.28
Molecular Formula: C10H20N2O2
InChi Key: FMLPQHJYUZTHQS-MRVPVSSYSA-N
SMILES: C[C@@H]1CN(CCN1)C(=O)OC(C)(C)C
Molecular Weight (g/mol) 
  • (2)
  • (1)
  • (2)
Quantity 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (4)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)