accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 163798-92-3

IUPAC Name: 4-[4-(bromomethyl)phenyl]-1,2,3-thiadiazole
Synonyms: 4-[4-(bromomethyl)phenyl]-1,2,3-thiadiazole
Molecular Weight (g/mol): 255.13
Molecular Formula: C9H7BrN2S
InChi Key: DGHQOPZIGDRUIT-UHFFFAOYSA-N
SMILES: BrCC1=CC=C(C=C1)C1=CSN=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results