accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3C

CAS RN 1638-26-2

IUPAC Name: 1,1-dimethylcyclopentane
Synonyms: 1,1-dimethylcyclopentane
Molecular Weight (g/mol): 98.19
Molecular Formula: C7H14
InChi Key: QWHNJUXXYKPLQM-UHFFFAOYSA-N
SMILES: CC1(C)CCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results