accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16387-70-5

IUPAC Name: octa-3,5-diyne
Synonyms: octa-3,5-diyne
Molecular Weight (g/mol): 106.17
Molecular Formula: C8H10
InChi Key: LILZEAJBVQOINI-UHFFFAOYSA-N
SMILES: CCC#CC#CCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results