Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Octadiyne 98.0+%, TCI America™
SDP

Catalog No. O01545ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O01545ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O01545ML Supplier TCI America Supplier No. O01545ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 16387-70-5
Molecular Formula C8H10
Molecular Weight (g/mol) 106.168
MDL Number MFCD00041633
InChI Key LILZEAJBVQOINI-UHFFFAOYSA-N
PubChem CID 140066
IUPAC Name octa-3,5-diyne
SMILES CCC#CC#CCC

Specifications

Boiling Point 164°C
Quantity 5 mL
UN Number 3295
Formula Weight 106.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,5-Octadiyne
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.