accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1639250-62-6

IUPAC Name: ethyl 2-amino-4-(4-cyanophenyl)thiophene-3-carboxylate
Synonyms: ethyl 2-amino-4-(4-cyanophenyl)thiophene-3-carboxylate
Molecular Weight (g/mol): 272.32
Molecular Formula: C14H12N2O2S
InChi Key: LVXVESIWKPHHIJ-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(N)SC=C1C1=CC=C(C=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results