accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C

CAS RN 1640-89-7

IUPAC Name: ethylcyclopentane
Synonyms: ethylcyclopentane
Molecular Weight (g/mol): 98.19
Molecular Formula: C7H14
InChi Key: IFTRQJLVEBNKJK-UHFFFAOYSA-N
SMILES: CCC1CCCC1
Molecular Weight (g/mol) 
  • (3)
  • (1)
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results