accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16405-35-9

IUPAC Name: platinum(2+) potassium ethene hydrate trichloride
Synonyms: zeise's salt
Molecular Weight (g/mol): 386.6
Molecular Formula: C2H6Cl3KOPt
InChi Key: DCEGWIMEFFONKJ-UHFFFAOYSA-K
SMILES: O.[Cl-].[Cl-].[Cl-].[K+].[Pt++].C=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results