Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium trichloro(ethylene)platinate(II) monohydrate, Pt 50.5% min

Catalog No. AA4402803
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4402803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4402803 Supplier Thermo Scientific Chemicals Supplier No. 04402803
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 16405-35-9 | C2H4Cl3KPt·H2O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16405-35-9
Molecular Formula C2H4Cl3KPt·H2O
MDL Number MFCD00150524

Specifications

Melting Point 220°C (decomposition)
Color Yellow
Physical Form Solid
Quantity 1 g
Linear Formula K[PtCl3(C2H4)]·H2O
Formula Weight 386.61 (368.59 Anhydrous)
Concentration or Composition (by Analyte or Components) Pt 50.5% min
Sensitivity Hygroscopic
Chemical Name or Material Potassium trichloro (ethylene)platinate (II) monohydrate
EINECSNumber 234-577-7
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.