accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16409-34-0

IUPAC Name: 2-[(4R)-4-[(1R,3aS,3bR,5aR,7R,9aS,9bS,11S,11aR)-7,11-dihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanamido]acetic acid sodium
Synonyms: glycodeoxycholic acid sodium
Molecular Weight (g/mol): 472.62
Molecular Formula: C26H43NNaO5
InChi Key: IIYSTUUVIASHBG-YEUHZSMFSA-N
SMILES: [Na].C[C@H](CCC(=O)NCC(O)=O)[C@H]1CC[C@H]2[C@@H]3CC[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results