Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Glycodeoxycholic Acid, Sodium Salt, ≥96%, Calbiochem™,

Catalog No. 3613115GM Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-131-15GM 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 36-131-15GM Supplier MilliporeSigma™ Supplier No. 3613115GM
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 16409-34-0
Molecular Formula C26H42NNaO5
Molecular Weight (g/mol) 471.614
InChI Key VMSNAUAEKXEYGP-GOIWIJIMSA-M
Synonym GDCA
PubChem CID 134129396
IUPAC Name sodium;2-[[(4R)-4-[(3R,5R,8R,9R,10S,12S,13R,14R,17S)-3,12-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]acetate
SMILES CC(CCC(=O)NCC(=O)[O-])C1CCC2C1(C(CC3C2CCC4C3(CCC(C4)O)C)O)C.[Na+]

Specifications

Color White
Quantity 5 g
Packaging Glass Bottle
Percent Purity ≥96% (TLC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.