accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHOOH3C

CAS RN 1641-17-4

IUPAC Name: 5-methoxy-2-(4-methylbenzoyl)phenol
Synonyms: mexenone benzophenone-10
Molecular Weight (g/mol): 242.27
Molecular Formula: C15H14O3
InChi Key: MJVGBKJNTFCUJM-UHFFFAOYSA-N
SMILES: COC1=CC=C(C(=O)C2=CC=C(C)C=C2)C(O)=C1
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results