accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSiOCH3OCH3OCH3

CAS RN 16415-12-6

IUPAC Name: hexadecyltrimethoxysilane
Synonyms: cetyl trimethoxysilane
Molecular Weight (g/mol): 346.63
Molecular Formula: C19H42O3Si
InChi Key: RSKGMYDENCAJEN-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCC[Si](OC)(OC)OC
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results