accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OCl(E/Z)

CAS RN 164220-45-5

IUPAC Name: 1-[2-(3-chlorophenyl)ethenyl]-3-methoxybenzene
Synonyms: 1-[2-(3-chlorophenyl)ethenyl]-3-methoxybenzene
Molecular Weight (g/mol): 244.72
Molecular Formula: C15H13ClO
InChi Key: DXHFJXLLHMOLGG-UHFFFAOYSA-N
SMILES: COC1=CC=CC(C=CC2=CC(Cl)=CC=C2)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results