Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-3'-methoxystilbene 97.0+%, TCI America™
SDP

Catalog No. C15931G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C15931G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C15931G Supplier TCI America Supplier No. C15931G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 164220-45-5
Molecular Formula C15H13ClO
Molecular Weight (g/mol) 244.72
MDL Number MFCD01861281
InChI Key DXHFJXLLHMOLGG-UHFFFAOYSA-N
PubChem CID 53421106
IUPAC Name 1-[2-(3-chlorophenyl)ethenyl]-3-methoxybenzene
SMILES COC1=CC=CC(C=CC2=CC(Cl)=CC=C2)=C1

Specifications

Melting Point 42°C
Color Yellow
Quantity 1 g
Formula Weight 244.72
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Chloro-3′-methoxystilbene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.