accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3O

CAS RN 16436-00-3

IUPAC Name: octyl 3-oxobutanoate
Synonyms: octyl 3-oxobutanoate
Molecular Weight (g/mol): 214.31
Molecular Formula: C12H22O3
InChi Key: IKYDDBGYKFPTGF-UHFFFAOYSA-N
SMILES: CCCCCCCCOC(=O)CC(C)=O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results