Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Octyl Acetoacetate 95.0+%, TCI America™
SDP

Catalog No. A091525ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A091525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A091525ML Supplier TCI America Supplier No. A091525ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 16436-00-3
Molecular Formula C12H22O3
Molecular Weight (g/mol) 214.31
MDL Number MFCD00059448
InChI Key IKYDDBGYKFPTGF-UHFFFAOYSA-N
Synonym Acetoacetic Acid n-Octyl Ester
PubChem CID 85418
IUPAC Name octyl 3-oxobutanoate
SMILES CCCCCCCCOC(=O)CC(C)=O

Specifications

Color Yellow
Boiling Point 198°C
Quantity 25 mL
Formula Weight 214.31
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material n-Octyl Acetoacetate
EINECSNumber (2)-1505
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.