accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSOCH3O

CAS RN 16437-69-7

IUPAC Name: (methylsulfanyl)methyl acetate
Synonyms: (methylsulfanyl)methyl acetate
Molecular Weight (g/mol): 120.17
Molecular Formula: C4H8O2S
InChi Key: LCUMNXCSNFGGGH-UHFFFAOYSA-N
SMILES: CSCOC(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results