Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylthiomethyl Acetate 95.0+%, TCI America™
SDP

Catalog No. A136825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A136825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A136825G Supplier TCI America Supplier No. A136825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 16437-69-7
Molecular Formula C4H8O2S
Molecular Weight (g/mol) 120.166
MDL Number MFCD00075528
InChI Key LCUMNXCSNFGGGH-UHFFFAOYSA-N
Synonym methylthiomethyl acetate, methylsulfanyl methyl acetate, acetic acid methylthiomethyl ester, methanol, methylthio-, acetate, methyl thiomethyl acetate, acmc-1c61t, methanol,1-methylthio-, 1-acetate
PubChem CID 167540
IUPAC Name methylsulfanylmethyl acetate
SMILES CC(=O)OCSC

Specifications

Color Colorless
Boiling Point 62°C
Quantity 25 g
UN Number 3272
Formula Weight 120.17
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Methylthiomethyl Acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.