accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1644-11-7

IUPAC Name: 1,1,1,2,2,3,3-heptafluoro-3-({1,1,1,2,3,3-hexafluoro-3-[(1,2,2-trifluoroethenyl)oxy]propan-2-yl}oxy)propane
Synonyms: 1,1,1,2,2,3,3-heptafluoro-3-({1,1,1,2,3,3-hexafluoro-3-[(1,2,2-trifluoroethenyl)oxy]propan-2-yl}oxy)propane
Molecular Weight (g/mol): 432.06
Molecular Formula: C8F16O2
InChi Key: RJBJXVAPYONTFE-UHFFFAOYNA-N
SMILES: FC(F)=C(F)OC(F)(F)C(F)(OC(F)(F)C(F)(F)C(F)(F)F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results