CAS RN 16455-98-4
IUPAC Name:
diantimony(3+) trioxalate
Synonyms:
diantimony(3+) trioxalate
Molecular Weight (g/mol):
507.57
Molecular Formula:
C6O12Sb2
InChi Key:
BMYPOELGNTXHPU-UHFFFAOYSA-H
SMILES:
[Sb+3].[Sb+3].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O
1
–
1
of
1
results
1
MP Biomedicals, Inc Antimony Oxalate, MP Biomedicals
CAS: 16455-98-4 Molecular Formula: C6O12Sb2 Molecular Weight (g/mol): 507.574 InChI Key: BMYPOELGNTXHPU-UHFFFAOYSA-H Synonym: antimony oxalate,diantimony 3+ trioxalate ion PubChem CID: 22141538 IUPAC Name: antimony(3+);oxalate SMILES: C(=O)(C(=O)[O-])[O-].C(=O)(C(=O)[O-])[O-].C(=O)(C(=O)[O-])[O-].[Sb+3].[Sb+3]