Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Antimony Oxalate, MP Biomedicals

Catalog No. ICN211051 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN211051 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN211051 Supplier MP Biomedicals, Inc Supplier No. 0521105180

Chemical Identifiers

CAS 16455-98-4
Molecular Formula C6O12Sb2
Molecular Weight (g/mol) 507.574
InChI Key BMYPOELGNTXHPU-UHFFFAOYSA-H
Synonym antimony oxalate, diantimony 3+ trioxalate ion
PubChem CID 22141538
IUPAC Name antimony(3+);oxalate
SMILES C(=O)(C(=O)[O-])[O-].C(=O)(C(=O)[O-])[O-].C(=O)(C(=O)[O-])[O-].[Sb+3].[Sb+3]

Specifications

Quantity 100 g
Chemical Name or Material Antimony Oxalate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.