accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHNOONHNSOOOSOOOSOOOSOOONaNaNaNa(E/Z)(E/Z)

CAS RN 164581-28-6

IUPAC Name: tetrasodium 4,5-dioxo-3,6-bis[2-(2-sulfonatophenyl)hydrazin-1-ylidene]-3,4,5,6-tetrahydronaphthalene-2,7-disulfonate
Synonyms: tetrasodium 4,5-dioxo-3,6-bis[2-(2-sulfonatophenyl)hydrazin-1-ylidene]naphthalene-2,7-disulfonate
Molecular Weight (g/mol): 776.55
Molecular Formula: C22H12N4Na4O14S4
InChi Key: OWVXGCUJHKOBMQ-UHFFFAOYSA-J
SMILES: [Na+].[Na+].[Na+].[Na+].[O-]S(=O)(=O)C1=CC=CC=C1NN=C1C(=O)C2=C(C=C1S([O-])(=O)=O)C=C(C(=NNC1=CC=CC=C1S([O-])(=O)=O)C2=O)S([O-])(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results