Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfonazo III Sodium Salt, Solstice

Catalog No. 60047700 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-047-700 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-047-700 Supplier Solstice Supplier No. 861705G
Add to Cart
Add to Cart

Titration Indicator (for SO4)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 164581-28-6
Molecular Formula C22H12N4Na4O14S4
Molecular Weight (g/mol) 776.551
MDL Number MFCD00003942
InChI Key OWVXGCUJHKOBMQ-SGLNGUFESA-J
PubChem CID 71299731
IUPAC Name tetrasodium;(3Z,6E)-4,5-dioxo-3,6-bis[(2-sulfonatophenyl)hydrazinylidene]naphthalene-2,7-disulfonate
SMILES C1=CC=C(C(=C1)NN=C2C(=CC3=C(C2=O)C(=O)C(=NNC4=CC=CC=C4S(=O)(=O)[O-])C(=C3)S(=O)(=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+].[Na+].[Na+]

Specifications

UN Number NONH for all modes of transport
Beilstein 1900148
Formula Weight 776.57g/mol
Packaging Poly bottle
Quantity 5 g
Physical Form Powder
Chemical Name or Material Sulfonazo III sodium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.