accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16498-20-7

IUPAC Name: 6-nitro-2,3-dihydro-1,4-benzodioxine
Synonyms: 6-nitro-2,3-dihydro-1,4-benzodioxine
Molecular Weight (g/mol): 181.15
Molecular Formula: C8H7NO4
InChi Key: MQSGCURTKWHBRX-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C2OCCOC2=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results