Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Ethylenedioxynitrobenzene, TCI America™
SDP

Catalog No. E05235G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E05235G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E05235G Supplier TCI America Supplier No. E05235G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 16498-20-7
Molecular Formula C8H7NO4
Molecular Weight (g/mol) 181.147
MDL Number MFCD00088689
InChI Key MQSGCURTKWHBRX-UHFFFAOYSA-N
Synonym 6-Nitro-1,4-benzodioxane
PubChem CID 85460
IUPAC Name 6-nitro-2,3-dihydro-1,4-benzodioxine
SMILES C1COC2=C(O1)C=CC(=C2)[N+](=O)[O-]

Specifications

Melting Point 121°C
Color Yellow
Quantity 5 g
Formula Weight 181.15
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Ethylenedioxynitrobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.