accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16499-57-3

IUPAC Name: 7-fluoro-1,4-dihydroquinazolin-4-one
Synonyms: 7-fluoro-1H-quinazolin-4-one
Molecular Weight (g/mol): 164.14
Molecular Formula: C8H5FN2O
InChi Key: KCORZHJVTZIZFD-UHFFFAOYSA-N
SMILES: FC1=CC=C2C(NC=NC2=O)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results