Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Fluoro-4-hydroxyquinazoline 98.0+%, TCI America™
SDP

Catalog No. F10291G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F10291G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F10291G Supplier TCI America Supplier No. F10291G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 16499-57-3
Molecular Formula C8H5FN2O
Molecular Weight (g/mol) 164.139
MDL Number MFCD05664874
InChI Key KCORZHJVTZIZFD-UHFFFAOYSA-N
Synonym 7-Fluoro-4-quinazolinone, 7-Fluoro-4-quinazolinol
PubChem CID 7018323
IUPAC Name 7-fluoro-1H-quinazolin-4-one
SMILES C1=CC2=C(C=C1F)NC=NC2=O

Specifications

Melting Point 248°C
Quantity 1 g
Formula Weight 164.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 7-Fluoro-4-hydroxyquinazoline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.