accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 165115-15-1

IUPAC Name: 2-(2-methyl-1,3-thiazol-4-yl)ethan-1-amine
Synonyms: 2-(2-methyl-1,3-thiazol-4-yl)ethanamine
Molecular Weight (g/mol): 142.22
Molecular Formula: C6H10N2S
InChi Key: GLEZGSKBHYTSAD-UHFFFAOYSA-N
SMILES: CC1=NC(CCN)=CS1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results