Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(2-Methyl-1,3-thiazol-4-yl)ethylamine, 97%, Thermo Scientific™

Catalog No. CC54613EA
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC54613EA 10 g Amber Glass Bottle
CC54613CB 250 mg Amber Glass Bottle
CC54613DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC54613EA Supplier Thermo Scientific Chemicals Supplier No. CC54613EA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 165115-15-1
Molecular Formula C6H10N2S
Molecular Weight (g/mol) 142.22
MDL Number MFCD03011601
InChI Key GLEZGSKBHYTSAD-UHFFFAOYSA-N
Synonym 2-2-methyl-1,3-thiazol-4-yl ethanamine, 2-2-methylthiazol-4-yl ethanamine, 2-2-methyl-1,3-thiazol-4-yl ethylamine, 4-thiazoleethanamine, 2-methyl, 2-2-methyl-1,3-thiazol-4-yl ethan-1-amine, 4-thiazoleethanamine,2-methyl, 2-2-methyl-1,3-thiazol-4-yl-1-ethanamine
PubChem CID 7536518
IUPAC Name 2-(2-methyl-1,3-thiazol-4-yl)ethanamine
SMILES CC1=NC(CCN)=CS1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 142.23
Percent Purity 97%
Chemical Name or Material 2-(2-Methyl-1, 3-thiazol-4-yl)ethylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.