accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1653-31-2

IUPAC Name: tridecan-2-ol
Synonyms: 2-tridecanol
Molecular Weight (g/mol): 200.37
Molecular Formula: C13H28O
InChi Key: HKOLRKVMHVYNGG-UHFFFAOYNA-N
SMILES: CCCCCCCCCCCC(C)O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)