Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Tridecanol, 97%

Catalog No. AAL0192606
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0192606 5 g
AAL0192603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0192606 Supplier Thermo Scientific Chemicals Supplier No. L0192606
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1653-31-2 | C13H28O | 200.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1653-31-2
Molecular Formula C13H28O
Molecular Weight (g/mol) 200.37
MDL Number MFCD00037601
InChI Key HKOLRKVMHVYNGG-UHFFFAOYNA-N
Synonym 2-tridecanol, 2-tridecyl alcohol, 2-hydroxytridecane, methylundecylcarbinol, 2-tridecanol, s, alcohols, c11-15-secondary, linear secondary c11-c15 alcohol, acmc-20m48e, acmc-1c00w
PubChem CID 15449
IUPAC Name tridecan-2-ol
SMILES CCCCCCCCCCCC(C)O

Specifications

Melting Point 23°C to 26°C
Density 0.83
Boiling Point 265°C
Refractive Index 1.443
Quantity 5 g
Beilstein 1720217
Formula Weight 200.37
Percent Purity 97%
Chemical Name or Material 2-Tridecanol
EINECSNumber 216-723-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.