accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOHOHOHOHNNN

CAS RN 165331-08-8

IUPAC Name: 2-(2-azidoethoxy)-6-(hydroxymethyl)oxane-3,4,5-triol
Synonyms: 2-(2-azidoethoxy)-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular Weight (g/mol): 249.22
Molecular Formula: C8H15N3O6
InChi Key: VGCOVRUENPCVTD-UHFFFAOYNA-N
SMILES: OCC1OC(OCCN=[N+]=[N-])C(O)C(O)C1O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results