Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Azidoethyl beta-D-Glucopyranoside 98.0+%, TCI America™
SDP

Catalog No. A22671G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A22671G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A22671G Supplier TCI America Supplier No. A22671G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 165331-08-8
Molecular Formula C8H15N3O6
Molecular Weight (g/mol) 249.223
InChI Key VGCOVRUENPCVTD-GWVFRZDISA-N
PubChem CID 53384348
IUPAC Name (2R,3S,4S,5S,6S)-2-(2-azidoethoxy)-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES C(COC1C(C(C(C(O1)CO)O)O)O)N=[N+]=[N-]

Specifications

Color Colorless
Quantity 1 g
Formula Weight 249.22
Percent Purity ≥98.0% (HPLC)
Physical Form Starch Syrup
Chemical Name or Material 2-Azidoethyl beta-D-Glucopyranoside
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.