accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1655-29-4

IUPAC Name: disodium naphthalene-1,5-disulfonate
Synonyms: disodium 1,5-naphthalenedisulfonate
Molecular Weight (g/mol): 332.25
Molecular Formula: C10H6Na2O6S2
InChi Key: YGSZNSDQUQYJCY-UHFFFAOYSA-L
SMILES: [Na+].[Na+].[O-]S(=O)(=O)C1=CC=CC2=C(C=CC=C12)S([O-])(=O)=O
Molecular Weight (g/mol) 
  • (3)
  • (1)
Percent Purity 
  • (2)
  • (1)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)