Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodium 1,5-naphthalenedisulfonate dibasic

Catalog No. 702401KG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-240-1KG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 70-240-1KG Supplier Sigma Aldrich Supplier No. 702401KG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Sodium 1,5-naphthalenedisulfonate dibasic

Specifications

CAS 1655-29-4
Molecular Formula C10H6Na2O6S2
Linear Formula C10H6Na2O6S2
MDL Number MFCD00064178
Quantity 250 mg
Synonym 1,5-Naphthalenedisulfonic acid disodium salt
Molecular Weight (g/mol) 332.26
Percent Purity ∼85% (T)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.