accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1655-52-3

IUPAC Name: (2S)-2-[(2,4-dinitrophenyl)amino]propanoic acid
Synonyms: N-(2,4-dinitrophenyl)alanine
Molecular Weight (g/mol): 255.19
Molecular Formula: C9H9N3O6
InChi Key: KKHLKORVTUUSBC-YFKPBYRVSA-N
SMILES: C[C@H](NC1=CC=C(C=C1[N+]([O-])=O)[N+]([O-])=O)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results