Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(2,4-Dinitrophenyl)-L-alanine 98.0+%, TCI America™
SDP

Catalog No. D10311G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D10311G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D10311G Supplier TCI America Supplier No. D10311G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1655-52-3
Molecular Formula C9H9N3O6
Molecular Weight (g/mol) 255.19
MDL Number MFCD00038106
InChI Key KKHLKORVTUUSBC-YFKPBYRVSA-N
Synonym N-Dnp-L-alanine, Dnp-Ala-OH
PubChem CID 5464517
IUPAC Name (2S)-2-[(2,4-dinitrophenyl)amino]propanoic acid
SMILES C[C@H](NC1=CC=C(C=C1[N+]([O-])=O)[N+]([O-])=O)C(O)=O

Specifications

Melting Point 179°C
Color Yellow
Quantity 1 g
UN Number 1325
Formula Weight 255.19
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(2,4-Dinitrophenyl)-L-alanine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.