CAS RN 16554-45-3
IUPAC Name:
2-methoxy-6-nitroaniline
Synonyms:
2-methoxy-6-nitroaniline
Molecular Weight (g/mol):
168.15
Molecular Formula:
C7H8N2O3
InChi Key:
NDKWDGCTUOOAPF-UHFFFAOYSA-N
SMILES:
COC1=CC=CC(=C1N)[N+]([O-])=O
1
–
1
of
1
results
1
Sigma Aldrich 2-Methoxy-6-nitroaniline
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.