accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16555-77-4

IUPAC Name: (2R)-2-hydroxy-2-(phenylformamido)acetate
Synonyms: (2R)-2-hydroxy-2-(phenylformamido)acetate
Molecular Weight (g/mol): 194.17
Molecular Formula: C9H8NO4
InChi Key: GCWCVCCEIQXUQU-MRVPVSSYSA-M
SMILES: O[C@@H](NC(=O)C1=CC=CC=C1)C([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results